3-Chloro-2-nitrotoluene ≥95%
Supplier: Matrix Scientific
072884-1G
072884-5G
102527-260EA
30.77
USD
102527-260
102527-262
3-Chloro-2-nitrotoluene ≥95%
3-Chloro-2-nitrotoluene
Hazard: Irritant
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol |
MDL Number:
MFCD04117977 CAS Number: 5367-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...