5-Hydroxy-2-tetralone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxy-3,4-dihydronaphthalen-2(1H)-one

072883-1G 072883-5G
102527-256EA 320 USD
102527-256 102527-258
5-Hydroxy-2-tetralone ≥95%
5-Hydroxy-2-tetralone

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD04038656
CAS Number: 35697-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR