Tropinone ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

072881-25G 072881-100G
102833-566EA 80 USD
102833-566 102833-568
Tropinone ≥95%
Tropinone

Hazard: Irritant

Formula: C₈H₁₃NO
MW: 139.2 g/mol
Boiling Pt: 227°C
Melting Pt: 40-43°C
MDL Number: MFCD00005549
CAS Number: 532-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR