6-Bromo-m-toluidine ≥95%
Supplier: Matrix Scientific
072880-10G
072880-100G
102527-242EA
143.08
USD
102527-242
102527-244
6-Bromo-m-toluidine ≥95%
6-Bromo-m-toluidine
Hazard: Irritant
Formula:
C₇H₈BrN MW: 186.05 g/mol |
MDL Number:
MFCD01806278 CAS Number: 53078-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...