5-Fluoro-2-iodobenzoic acid ≥95%
Supplier: Matrix Scientific
072878-1G
072878-5G
072878-10G
102527-232EA
364.62
USD
102527-232
102527-234
102527-236
5-Fluoro-2-iodobenzoic acid ≥95%
5-Fluoro-2-iodobenzoic acid
Hazard: Irritant
Formula:
C₇H₄FIO₂ MW: 266.01 g/mol |
MDL Number:
MFCD00837316 CAS Number: 52548-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...