6-Methoxy-2-methylpyridin-3-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Methoxy-2-methylpyridin-3-amine, 3-Amino-6-methoxy-2-methylpyridine, 3-Amino-6-methoxy-2-picoline
072876-1G
072876-5G
102527-222EA
69.23
USD
102527-222
102527-224
6-Methoxy-2-methylpyridin-3-amine ≥95%
6-Methoxy-2-methylpyridin-3-amine
Hazard: Irritant
Formula:
C₇H₁₀N₂O MW: 138.1671 g/mol |
MDL Number:
MFCD03095090 CAS Number: 52090-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...