trans-4-Aminocyclohexanol hydrochloride ≥95%

Supplier: Matrix Scientific

072875-100G 072875-500G
102527-218EA 110.77 USD
102527-218 102527-220
trans-4-Aminocyclohexanol hydrochloride ≥95%
trans-4-Aminocyclohexanol hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNO
MDL Number: MFCD00012566
CAS Number: 50910-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR