2,4,6-Trichloropyrimidine-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4,6-Trichloro-5-formylpyrimidine, 2,4,6-Trichloro-5-pyrimidinecarboxaldehyde

072873-5G 072873-25G
102527-208EA 60 USD
102527-208 102663-910
2,4,6-Trichloropyrimidine-5-carbaldehyde ≥95%
2,4,6-Trichloropyrimidine-5-carbaldehyde

Hazard: Toxic/Refrigerate

Formula: C₅HCl₃N₂O
MW: 211.43 g/mol
Melting Pt: 131…132 °C
MDL Number: MFCD02257700
CAS Number: 50270-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR