3-Pyridineacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (Pyridin-3-yl)acetic acid

072872-5G 072872-100G
102527-200EA 227.69 USD
102527-200 102527-204
3-Pyridineacetic acid ≥95%
3-Pyridineacetic acid

Hazard: Irritant

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 144…146 °C
CAS Number: 501-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR