2-Bromo-4,6-dimethylpyridine ≥95%

Supplier: Matrix Scientific

072868-1G 072868-5G
102527-180EA 138.46 USD
102527-180 102527-182
2-Bromo-4,6-dimethylpyridine ≥95%
2-Bromo-4,6-dimethylpyridine

Hazard: Irritant

Formula: C₇H₈BrN
MDL Number: MFCD00082591
CAS Number: 4926-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR