4-Hydroxypyrimidine-5-carbonitrile ≥95%

Supplier: Matrix Scientific

072867-250MG 072867-1G 072867-5G
102527-174EA 400 USD
102527-174 102527-176 102527-178
4-Hydroxypyrimidine-5-carbonitrile ≥95%
4-Hydroxypyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₅H₃N₃O
CAS Number: 4774-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR