(5-Methylpyridin-2-yl)methanamine ≥95%

Supplier: Matrix Scientific

072865-250MG 072865-1G 072865-5G
102527-164EA 483.85 USD
102527-164 102527-166 102527-168
(5-Methylpyridin-2-yl)methanamine ≥95%
(5-Methylpyridin-2-yl)methanamine

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.1677 g/mol
MDL Number: MFCD08752541
CAS Number: 45715-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR