3-Fluoro-4-nitroanisole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-methoxy-1-nitrobenzene

072863-1G 072863-5G 072863-25G
102527-152EA 15.38 USD
102527-152 102527-154 102833-560
3-Fluoro-4-nitroanisole ≥95%
3-Fluoro-4-nitroanisole

Hazard: Irritant

Formula: C₇H₆FNO₃
MW: 171.13 g/mol
Melting Pt: 56…58 °C
MDL Number: MFCD04115632
CAS Number: 446-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR