5-Methylpyridin-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-5-methylpyridine, 3-Hydroxy-5-picoline

072854-1G 072854-5G
102527-104EA 146.15 USD
102527-104 102527-106
5-Methylpyridin-3-ol ≥95%
5-Methylpyridin-3-ol

Hazard: Irritant

Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00661297
CAS Number: 42732-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR