5-Hydroxy-2-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

072853-25G 072853-100G
102833-556EA 143.08 USD
102833-556 102833-558
5-Hydroxy-2-nitrobenzaldehyde ≥95%
5-Hydroxy-2-nitrobenzaldehyde

Hazard: Irritant/Air Sensitive/Store in Freezer

Formula: C₇H₅NO₄
Melting Pt: 166-168°C
MDL Number: MFCD00007332
CAS Number: 42454-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR