5-Chlorothiazol-2-amine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (5-Chlorothiazol-2-yl)amine hydrochloride, 2-Amino-5-chlorothiazole hydrochloride

072851-1G 072851-5G
102527-090EA 150.77 USD
102527-090 102527-092
5-Chlorothiazol-2-amine hydrochloride ≥95%
5-Chlorothiazol-2-amine hydrochloride

Hazard: Irritant

Formula: C3H4Cl2N2S
MW: 171.05 g/mol
MDL Number: MFCD00039006
CAS Number: 55506-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR