3-Methylpyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

072849-1G 072849-5G
102527-076EA 70.77 USD
102527-076 102527-078
3-Methylpyrazine-2-carboxylic acid ≥95%
3-Methylpyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C6H6N2O2
MW: 138.1200 g/mol
MDL Number: MFCD08705765
CAS Number: 41110-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR