4-Chloro-2-hydroxypyridine ≥95%

Supplier: Matrix Scientific

072848-1G 072848-5G 072848-10G
102527-070EA 61.54 USD
102527-070 102527-072 102527-074
4-Chloro-2-hydroxypyridine ≥95%
4-Chloro-2-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄ClNO
MW: 129.55 g/mol
MDL Number: MFCD03093022
CAS Number: 40673-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR