4-Bromo-3-(trifluoromethyl)anisole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-methoxybenzotrifluoride, 4-Bromo-3-trifluoromethylanisole

072845-1G 072845-5G 072845-10G
102527-054EA 30.77 USD
102527-054 102527-056 102527-058
4-Bromo-3-(trifluoromethyl)anisole ≥95%
4-Bromo-3-(trifluoromethyl)anisole

Hazard: Irritant

Formula: C₈H₆BrF₃O
MW: 255.04 g/mol
MDL Number: MFCD08059527
CAS Number: 400-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR