2-Chloro-3-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

072842-5G 072842-25G
102527-040EA 107.69 USD
102527-040 102551-812
2-Chloro-3-nitrobenzoic acid ≥95%
2-Chloro-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₄ClNO₄
MDL Number: MFCD00007069
CAS Number: 3970-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR