4,5,6,7-Tetrahydro-1-phenyl-1H-pyrazolo[4,3-c]pyridine ≥95%

Supplier: Matrix Scientific

072841-250MG 072841-1G
102527-034EA 740 USD
102527-034 102527-036
4,5,6,7-Tetrahydro-1-phenyl-1H-pyrazolo[4,3-c]pyridine ≥95%
4,5,6,7-Tetrahydro-1-phenyl-1H-pyrazolo[4,3-c]pyridine

Hazard: Irritant

Formula: C₁₂H₁₃N₃
MW: 199.25 g/mol
MDL Number: MFCD11044739
CAS Number: 396133-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR