3-Chloro-4-nitrobenzoic acid ≥95%
Supplier: Matrix Scientific
072840-5G
072840-100G
102527-028EA
30.77
USD
102527-028
102527-032
3-Chloro-4-nitrobenzoic acid ≥95%
3-Chloro-4-nitrobenzoic acid
Hazard: Irritant
Formula:
C₇H₄ClNO₄ MW: 201.57 g/mol |
MDL Number:
MFCD01632172 CAS Number: 39608-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...