5-Chloro-1,2,4-thiadiazole ≥95%

Supplier: Matrix Scientific

072835-1G 072835-5G
102527-000EA 153.85 USD
102527-000 102527-002
5-Chloro-1,2,4-thiadiazole ≥95%
5-Chloro-1,2,4-thiadiazole

Hazard: Irritant

Formula: C₂HClN₂S
MDL Number: MFCD09743974
CAS Number: 38362-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR