3H-Spiro[2-benzofuran-1,4'-piperidine] ≥95%

Supplier: Matrix Scientific

072834-250MG 072834-1G 072834-5G
102526-994EA 381.12 USD
102526-994 102526-996 102526-998
3H-Spiro[2-benzofuran-1,4'-piperidine] ≥95%
3H-Spiro[2-benzofuran-1,4'-piperidine]

Hazard: Irritant

Formula: C12H15NO
CAS Number: 38309-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR