5-Acetyl-2-methylpyridine ≥95%

Supplier: Matrix Scientific

072831-1G 072831-5G
102526-978EA 221.54 USD
102526-978 102526-980
5-Acetyl-2-methylpyridine ≥95%
5-Acetyl-2-methylpyridine

Hazard: Irritant

Formula: C₈H₉NO
MDL Number: MFCD00128113
CAS Number: 36357-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR