2-Methylthiazole ≥95%

Supplier: Matrix Scientific

072829-5G 072829-10G 072829-100G
102526-970EA 61.54 USD
102526-970 102526-972 102526-974
2-Methylthiazole ≥95%
2-Methylthiazole

Hazard: Irritant

Formula: C₄H₅NS
MW: 99.16 g/mol
MDL Number: MFCD00053144
CAS Number: 3581-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR