N-Ethyl-4-hydroxypiperidine ≥95%

Supplier: Matrix Scientific

072827-1G 072827-5G
102526-960EA 533.85 USD
102526-960 102526-962
N-Ethyl-4-hydroxypiperidine ≥95%
N-Ethyl-4-hydroxypiperidine

Hazard: Irritant

Formula: C₇H₁₅NO
MW: 129.2001 g/mol
MDL Number: MFCD00006509
CAS Number: 3518-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR