(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
(3S)-1,2,3,4-Tetrahydro-3-isoquinolinecarbocylic acid, (S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
072826-5G
072826-10G
072826-100G
102526-954EA
143.08
USD
102526-954
102526-956
102526-958
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥95%
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₁NO₂ MW: 177.20 g/mol |
MDL Number:
MFCD00144533 CAS Number: 74163-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...