4-Chloro-2-fluoropyridine ≥95%

Supplier: Matrix Scientific

072825-1G 072825-5G
102526-950EA 20 USD
102526-950 102526-952
4-Chloro-2-fluoropyridine ≥95%
4-Chloro-2-fluoropyridine

Hazard: Irritant

Formula: C₅H₃ClFN
MDL Number: MFCD04112503
CAS Number: 34941-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR