3,5-Dibromo-2,6-dimethylpyridine ≥95%

Supplier: Matrix Scientific

072819-250MG 072819-1G 072819-5G
102526-916EA 400 USD
102526-916 102526-918 102526-920
3,5-Dibromo-2,6-dimethylpyridine ≥95%
3,5-Dibromo-2,6-dimethylpyridine

Hazard: Irritant

Formula: C7H7Br2N
MDL Number: MFCD00234282
CAS Number: 3430-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR