5-Methoxy-1-tetralone ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Methoxy-3,4-dihydronaphthalenone
072817-1G
072817-5G
072817-10G
102526-904EA
55.38
USD
102526-904
102526-906
102526-908
5-Methoxy-1-tetralone ≥95%
5-Methoxy-1-tetralone
Hazard: Irritant
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol |
MDL Number:
MFCD00001692 CAS Number: 33892-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...