6-Hydroxy-1,2,3,4-tetrahydroquinoline ≥95%
Supplier: Matrix Scientific
Synonyms:
1,2,3,4-Tetrahydroquinolin-6-ol
072816-250MG
072816-1G
072816-5G
102526-898EA
450.42
USD
102526-898
102526-900
102526-902
6-Hydroxy-1,2,3,4-tetrahydroquinoline ≥95%
6-Hydroxy-1,2,3,4-tetrahydroquinoline
Hazard: Irritant
Formula:
C₉H₁₁NO MW: 149.1897 g/mol |
MDL Number:
MFCD00800243 CAS Number: 3373-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...