5-Methoxy-2-tetralone ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Dihydro-5-methoxy-2(1H)-naphthalenone

072814-1G 072814-5G 072814-10G
102526-886EA 143.08 USD
102526-886 102526-888 102526-890
5-Methoxy-2-tetralone ≥95%
5-Methoxy-2-tetralone

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD00064958
CAS Number: 32940-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR