5,6-Diamino-2,4-dihydroxypyrimidine ≥95%

Supplier: Matrix Scientific

072813-1G 072813-5G
102526-882EA 427.69 USD
102526-882 102526-884
5,6-Diamino-2,4-dihydroxypyrimidine ≥95%
5,6-Diaminopyrimidine-2,4-diol

Hazard: Irritant

Formula: C₄H₆N₄O₂
MDL Number: MFCD00016632
CAS Number: 3240-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR