4-Methoxy-3-nitropyridine ≥95%

Supplier: Matrix Scientific

072809-5G 072809-25G
102526-860EA 96.92 USD
102526-860 102551-806
4-Methoxy-3-nitropyridine ≥95%
4-Methoxy-3-nitropyridine

Hazard: Irritant

Formula: C₆H₆N₂O₃
MDL Number: MFCD00209661
CAS Number: 31872-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR