6-Nitro-m-toluic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Nitro-5-methylbenzoic acid, 5-Methyl-2-nitrobenzoic acid

072806-10G 072806-100G
102526-842EA 178.46 USD
102526-842 102526-844
6-Nitro-m-toluic acid ≥95%
6-Nitro-m-toluic acid

Hazard: Irritant

Formula: C₈H₇NO₄
MDL Number: MFCD00007368
CAS Number: 3113-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR