Ethyl-3-methyl-4-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-methyl-4-nitrobenzoate

072805-10G 072805-100G
102526-838EA 107.69 USD
102526-838 102526-840
Ethyl-3-methyl-4-nitrobenzoate ≥95%
Ethyl-3-methyl-4-nitrobenzoate

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
MW: 209.20 g/mol
MDL Number: MFCD00115804
CAS Number: 30650-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR