4-(Hydroxymethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Carboxybenzylalcohol, HMBA Linker, HMBA

072802-5G 072802-100G
102526-820EA 56.92 USD
102526-820 102526-824
4-(Hydroxymethyl)benzoic acid ≥95%
4-(Hydroxymethyl)benzoic acid

Hazard: Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 179…183 °C
MDL Number: MFCD00017598
CAS Number: 3006-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR