Methyl-5-bromopicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromopyridine-2-carboxylic acid methyl ester, Methyl 5-bromopyridine-2-carboxylate, Methyl 5-bromopicolinate

072799-5G 072799-25G 072799-100G
102526-804EA 126.15 USD
102526-804 102663-900 102663-902
Methyl-5-bromopicolinate ≥95%
Methyl 5-bromopicolinate

Hazard: Irritant

Formula: C₇H₆BrNO₂
Melting Pt: 99-103°C
MDL Number: MFCD04112493
CAS Number: 29682-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR