tert-Butyl-8-azabicyclo[3.2.1]octan-3-ylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 8-azabicyclo[3.2.1]octan-3-ylcarbamate

072797-250MG 072797-1G
102526-794EA 710.77 USD
102526-794 102526-796
tert-Butyl-8-azabicyclo[3.2.1]octan-3-ylcarbamate ≥95%
tert-Butyl-8-azabicyclo[3.2.1]octan-3-ylcarbamate

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MW: 226.3200 g/mol
MDL Number: MFCD11045410
CAS Number: 287114-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR