5-Methoxyisobenzofuran-1,3-dione ≥95%

Supplier: Matrix Scientific

072796-1G 072796-5G
102526-790EA 710.77 USD
102526-790 102526-792
5-Methoxyisobenzofuran-1,3-dione ≥95%
5-Methoxyisobenzofuran-1,3-dione
Formula: C9H6O4
MDL Number: MFCD02670942
CAS Number: 28281-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR