trans-2,5-Dimethylpiperazine ≥95%

Supplier: Matrix Scientific

072795-10G 072795-100G
102526-786EA 126.15 USD
102526-786 102526-788
trans-2,5-Dimethylpiperazine ≥95%
trans-2,5-Dimethylpiperazine

Hazard: Irritant

Formula: C₆H₁₄N₂
MDL Number: MFCD00070516
CAS Number: 2815-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR