2,5-Dimethoxybenzoic acid ≥95%

Supplier: Matrix Scientific

072794-5G 072794-10G 072794-100G
102526-780EA 107.69 USD
102526-780 102526-782 102526-784
2,5-Dimethoxybenzoic acid ≥95%
2,5-Dimethoxybenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00002436
CAS Number: 2785-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR