1H-Pyrazole-4-boronic acid pinacol ester ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
072791-1G
072791-5G
102526-766EA
30.77
USD
102526-766
102526-768
1H-Pyrazole-4-boronic acid pinacol ester ≥95%
1H-Pyrazole-4-boronic acid pinacol ester
Hazard: Irritant/Store in Freezer
Formula:
C₉H₁₅BN₂O₂ MW: 194.039 g/mol Melting Pt: 145-149°C |
MDL Number:
MFCD03453063 CAS Number: 269410-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...