3,11-Dichloro-6-methyl-6,11-dihydrodibenzo[c,f][1,2]thiazepine 5,5-dioxide ≥95%

Supplier: Matrix Scientific

072788-1G 072788-5G 072788-10G
102526-748EA 178.46 USD
102526-748 102526-750 102526-752
3,11-Dichloro-6-methyl-6,11-dihydrodibenzo[c,f][1,2]thiazepine 5,5-dioxide ≥95%
3,11-Dichloro-6-methyl-6,11-dihydrodibenzo[c,f][1,2]thiazepine 5,5-dioxide

Hazard: Irritant

Formula: C14H11Cl2NO2S
MW: 328.213 g/mol
MDL Number: MFCD09029354
CAS Number: 26638-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR