Ethyl-6-bromonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-bromo-5-pyridinecarboxylate, Ethyl 6-bromonicotinate, 6-Bromonicotinic acid ethyl ester

072786-1G 072786-5G 072786-10G
102526-736EA 355.38 USD
102526-736 102526-738 102526-740
Ethyl-6-bromonicotinate ≥95%
Ethyl-6-bromonicotinate

Hazard: Irritant

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
MDL Number: MFCD11111853
CAS Number: 132334-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR