1-Fluoro-3-ethynylbenzene ≥95%

Supplier: Matrix Scientific

Synonyms: (3-Fluorophenyl)ethyne, 3-Fluorophenylacetylene, (3-Fluorophenyl)acetylene, 1-Ethynyl-3-fluorobenzene

072783-5G 072783-100G
102526-720EA 73.85 USD
102526-720 102526-724
1-Fluoro-3-ethynylbenzene ≥95%
1-Fluoro-3-ethynylbenzene

Hazard: Irritant

Formula: C₈H₅F
MW: 120.13 g/mol
MDL Number: MFCD03093798
CAS Number: 2561-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR