4-Bromo-3-fluoropyridine ≥95%
Supplier: Matrix Scientific
072782-1G
072782-5G
102526-716EA
276.92
USD
102526-716
102526-718
4-Bromo-3-fluoropyridine ≥95%
4-Bromo-3-fluoropyridine
Hazard: Irritant
Formula:
C₅H₃BrFN |
MDL Number:
MFCD04113266 CAS Number: 2546-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...