4-Methyloxazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

072781-1G 072781-5G 072781-10G
102526-710EA 284.62 USD
102526-710 102526-712 102526-714
4-Methyloxazole-5-carboxylic acid ≥95%
4-Methyloxazole-5-carboxylic acid

Hazard: Irritant

Formula: C₅H₅NO₃
MW: 127.1000 g/mol
MDL Number: MFCD01571070
CAS Number: 2510-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR