3-Methoxy-4-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

072780-1G 072780-5G 072780-10G
102526-704EA 161.54 USD
102526-704 102526-706 102526-708
3-Methoxy-4-methylbenzaldehyde ≥95%
3-Methoxy-4-methylbenzaldehyde

Hazard: Irritant

Formula: C9H10O2
MW: 150.1700 g/mol
MDL Number: MFCD04114050
CAS Number: 24973-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR